Anthracene-D10, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Manufacturer:  Spex Certiprep Llc S291*
Catalog No. S-291
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
Anthracene-d101719-06-81000μg/mL
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
2mL amber prescored glass ampule
1mL
GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
MeCl
Chemical Identifiers
0.0
icon-pdf.png
SDS
Documents
Product Certifications
Product intended for laboratory use only