missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Anthracene, 99%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific 104860025

 View more versions of this product

Catalog No. AC104860025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Anthracene
120-12-7
215.0°C to 218.0°C
121°C
C14H10
MFCD00001240
05,657
15,674
anthracen, anthracen german, anthracene, pure, anthracin, anthrazen, bis-alkylamino, ccris 767, green oil, paranaphthalene, tetra olive n2g
MWPLVEDNUUSJAV-UHFFFAOYSA-N
anthracene
8418
178.23
99%
99%
Yellow
340.0°C
98.5% min. (GC)
Authentic
Plastic bottle
12,290; 15,99; 16,13
2.5kg
Solubility in water: insoluble. Other solubilities: 1g/67mL alcohol - 1g/70mL methanol, 1g/200mL ether - 1g/62mL benzene, 1g/85mL chloroform - 1g/31mL carbon disulfide, 1g/86mL ccl4 - 1g/125mL toluene
C1=CC2=CC3=CC=CC=C3C=C2C=C1
178.23
CHEBI:35298
Crystalline Powder

Chemical Identifiers

120-12-7
178.23
MWPLVEDNUUSJAV-UHFFFAOYSA-N
8418
anthracene
C14H10
MFCD00001240
anthracen, anthracen german, anthracene, pure, anthracin, anthrazen, bis-alkylamino, ccris 767, green oil, paranaphthalene, tetra olive n2g
CHEBI:35298
C1=CC2=CC3=CC=CC=C3C=C2C=C1
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Warning

EINECSNumber : 204-371-1

RTECSNumber : CA9350000

TSCA : TSCA

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only