Learn More
Anthracene, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1020322
Description
Anthracene is used in the production of the red dyealizarin and other dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAnthracene is used in the production of the red dyealizarin and other dyes.
Solubility
Soluble in alcohol, methanol (1.8 g/100 g), hexane (0.37 g/100 g). Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
Specifications
Anthracene | |
214°C to 218°C | |
340°C to 342°C | |
C14H10 | |
100 g | |
1905429 | |
paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767 | |
MWPLVEDNUUSJAV-UHFFFAOYSA-N | |
anthracene | |
8418 | |
178.23 |
120-12-7 | |
1.25 | |
121°C (249°F) | |
MFCD00001240 | |
UN3077 | |
14,682 | |
Soluble in alcohol,methanol (1.8g/100 g),hexane (0.37g/100 g). Insoluble in water. | |
C1=CC2=CC3=CC=CC=C3C=C2C=C1 | |
178.23 | |
CHEBI:35298 | |
99% |
Chemical Identifiers
120-12-7 | |
178.23 | |
MWPLVEDNUUSJAV-UHFFFAOYSA-N | |
8418 | |
anthracene |
C14H10 | |
MFCD00001240 | |
paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767 | |
CHEBI:35298 | |
C1=CC2=CC3=CC=CC=C3C=C2C=C1 |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319-H500
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 204-371-1
RTECSNumber : CA9350000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only