Please login to your online account to display your discounted pricing

Alfa Aesar™ Anthracene, 97%

Chemical Identifiers

Chemical Name or Material Anthracene
CAS 120-12-7
Molecular Formula C14H10
Formula Weight 178.23
MDL Number MFCD00001240
UN Number UN3077
Beilstein 1905429
Merck Index 14,682
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA172610B

  SDS
alfa aesar™
A172610B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA172610I

  SDS
alfa aesar™
A172610I
5000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1726130

  SDS
alfa aesar™
A1726130
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 178.23
Molecular Formula C14H10
Merck Index 14,682
Health Hazard 2 GHS H Statement
H317
May cause an allergic skin reaction.
Boiling Point 340° to 342°C
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number CA9350000
UN Number UN3077
Chemical Name or Material Anthracene
Quantity 1000g
Melting Point 213° to 216°C
Beilstein 1905429
TSCA TSCA
Flash Point 121°C (249°F)
MDL Number MFCD00001240
Health Hazard 3 GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 120-12-7
Assay 97%
Health Hazard 1 Warning

Applications
Anthracene is used as a source of dyestuffs and involved in the preparation of red dye alizarin. It is an organic semiconductor used as a scintillator for detectors of high energy photons, electrons and alfa particles. It is doped with polyvinyltoluene to prepare plastic scintillator. Further, it is used in the preparation of anthraquinone, wood preservatives, coating materials and pigments. In addition to this, it is used to prepare 1-hydroxyanthracene, 2-hydroxyanthracene and in 9,10-dihydroxyanthracene.

Solubility
Soluble in methanol, benzene, chloroform, ether, carbon disulfide, carbon tetrachloride and toluene. Slightly soluble in acetone. Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents and hypochlorites.

Safety and Handling

Warning

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H317
May cause an allergic skin reaction.