Please login to your online account to display your discounted pricing

Alfa Aesar™ Anthracene, 90+%

Chemical Identifiers

Chemical Name or Material Anthracene
CAS 120-12-7
Formula Weight 178.23
MDL Number MFCD00001240
UN Number UN3077
Beilstein 1905429
Merck Index 14,682
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3232036

  SDS
alfa aesar™
3232036
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA32320A3

  SDS
alfa aesar™
32320A3
2kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 178.23
Chemical Name or Material Anthracene
TSCA TSCA
Density 1.250g/mL
Physical Form Crystalline
Assay ≥90%
MDL Number MFCD00001240
CAS 120-12-7
UN Number UN3077
Beilstein 1905429
Merck Index 14,682
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H317
May cause an allergic skin reaction.
Boiling Point 340° to 342°C
Melting Point 210° to 215°C
Quantity 500g
Health Hazard 3 GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 121° (249°F)
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number CA9350000
Safety and Handling

Warning

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H317
May cause an allergic skin reaction.