missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Anthracene, 90+%

Manufacturer:  Alfa Aesar™ 32320A3

 View more versions of this product

Catalog No. AA32320A3



Specifications

Specifications

Anthracene
1.250g/mL
C14H10
UN3077
14,682
C1=CC=C2C=C3C=CC=CC3=CC2=C1
178.234
CHEBI:35298
Crystalline
210°C to 215°C
121° (249°F)
120-12-7
≥90%
MFCD00001240
1905429
MWPLVEDNUUSJAV-UHFFFAOYSA-N
anthracene
8418
178.23
340°C to 342°C
2kg

Chemical Identifiers

120-12-7
178.234
MWPLVEDNUUSJAV-UHFFFAOYSA-N
CHEBI:35298
C1=CC=C2C=C3C=CC=CC3=CC2=C1
C14H10
MFCD00001240
8418
anthracene
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : CA9350000

TSCA : TSCA

SDS Documents