Please login to your online account to display your discounted pricing

Alfa Aesar™ Anthracene-9-boronic acid, 99%

Chemical Identifiers

Chemical Name or Material Anthracene-9-boronic acid
CAS 100622-34-2
Molecular Formula C14H11BO2
Formula Weight 222.06
MDL Number MFCD03425925
UN Number UN2811
Beilstein 3301031
Synonym 9-Anthrylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL19630MC

 
alfa aesar™
L19630MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL19630ME

 
alfa aesar™
L19630ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 222.06
Chemical Name or Material Anthracene-9-boronic acid
Assay 99%
Molecular Formula C14H11BO2
Synonym 9-Anthrylboronic acid
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD03425925
CAS 100622-34-2
UN Number UN2811
Beilstein 3301031
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈215°C
Quantity 100mg
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.