missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Anthracene-9-boronic acid, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC432812500

 View more versions of this product

Catalog No. AC432812500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Anthracene-9-boronic acid
Glass bottle
250mg
97%
MFCD03425925
VHHDLIWHHXBLBK-UHFFFAOYSA-N
anthracen-9-ylboronic acid
15510213
Crystalline Solid
Authentic
Tan to White
100622-34-2
C14H11BO2
9-anthraceneboronic acid, anthracene-9-boronic acid, 9-anthracenylboronic acid, 9-anthrylboronic acid, 9-anthracene boronic acid, boronic acid, 9-anthracenyl, 9-boronoanthracene, 9-borono-anthracene, zlchem 414, pubchem7757
B(C1=C2C=CC=CC2=CC3=CC=CC=C13)(O)O
222.05
222.05
≥96.0%

Chemical Identifiers

100622-34-2
222.05
VHHDLIWHHXBLBK-UHFFFAOYSA-N
15510213
B(C1=C2C=CC=CC2=CC3=CC=CC=C13)(O)O
C14H11BO2
MFCD03425925
9-anthraceneboronic acid, anthracene-9-boronic acid, 9-anthracenylboronic acid, 9-anthrylboronic acid, 9-anthracene boronic acid, boronic acid, 9-anthracenyl, 9-boronoanthracene, 9-borono-anthracene, zlchem 414, pubchem7757
anthracen-9-ylboronic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents