missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Anthracene-9,10-diboronic acid bis(pinacol) ester, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5192106

 View more versions of this product

Catalog No. AAH5192106



Description

Description

It is used synthesis, structure and electrochemistry of mono- and dinuclear osmium n,n-di- and tetraphenylbipyridyls and extended bipyridyls. It is also used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used synthesis, structure and electrochemistry of mono- and dinuclear osmium n,n′-di- and tetraphenylbipyridyls and extended bipyridyls. It is also used as a primary and secondary intermediate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Anthracene-9,10-diboronic acid bis(pinacol) ester
C26H32B2O4
5 g
ZLXSWNVYGSZXOP-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-[10-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anthracen-9-yl]-1,3,2-dioxaborolane
57415691
95%
863992-56-7
MFCD16294538
9,10-anthracenediboronic acid bis pinacol ester, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anthracene, 4,4,5,5-tetramethyl-2-10-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anthracen-9-yl-1,3,2-dioxaborolane, amtb237, anthracene-9,10-diboronic acid bis pinacol ester, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolanyl anthracene, 9,10-bis 4,4,5,5-tetramethyl 1.3.2 dioxaborolan-2-yl anthracene, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl anthracene, 1,3,2-dioxaborolane, 2,2'-9,10-anthracenediyl bis 4,4,5,5-tetramethyl
CC1(C)OB(OC1(C)C)C1=C2C=CC=CC2=C(B2OC(C)(C)C(C)(C)O2)C2=CC=CC=C12
430.16
430.16

Chemical Identifiers

863992-56-7
430.16
ZLXSWNVYGSZXOP-UHFFFAOYSA-N
57415691
CC1(C)OB(OC1(C)C)C1=C2C=CC=CC2=C(B2OC(C)(C)C(C)(C)O2)C2=CC=CC=C12
C26H32B2O4
MFCD16294538
9,10-anthracenediboronic acid bis pinacol ester, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anthracene, 4,4,5,5-tetramethyl-2-10-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anthracen-9-yl-1,3,2-dioxaborolane, amtb237, anthracene-9,10-diboronic acid bis pinacol ester, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolanyl anthracene, 9,10-bis 4,4,5,5-tetramethyl 1.3.2 dioxaborolan-2-yl anthracene, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl anthracene, 1,3,2-dioxaborolane, 2,2'-9,10-anthracenediyl bis 4,4,5,5-tetramethyl
4,4,5,5-tetramethyl-2-[10-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anthracen-9-yl]-1,3,2-dioxaborolane
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only