Please login to your online account to display your discounted pricing

Antalarmin hydrochloride, Tocris

CRF1 antagonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material Antalarmin hydrochloride
Assay Percent Range >99%
Molecular Formula C24H34N4·HCl
Formula Weight 415.01
Synonym N-Butyl-N-ethyl-2,5,6-trimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

277810

 
r & d systems
277810
10mg Each for $252.39

277850

 
r & d systems
277850
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 415.01
Quantity 10mg
Molecular Formula C24H34N4·HCl
CAS 220953-69-5
Synonym N-Butyl-N-ethyl-2,5,6-trimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride
Chemical Name or Material Antalarmin hydrochloride
Assay Percent Range >99%
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C