Please login to your online account to display your discounted pricing

Alfa Aesar™ Antagonist G

For cellular and molecular biology applications

Specifications

Product Type Antagonist G
Molecular Weight 951.20
Storage Requirements -30° to -10°C
Sequence Arg-D-Trp-N-Me-Phe-D-Trp-Leu-Met-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66045MCR

 
alfa aesar™
J66045MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66045EXC

 
alfa aesar™
J66045EXC
2.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Antagonist G
Molecular Weight 951.20
Storage Requirements -30° to -10°C
Sequence Arg-D-Trp-N-Me-Phe-D-Trp-Leu-Met-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC