Please login to your online account to display your discounted pricing
Ansell™ Vantage™ Cut-Resistant Gloves
Superior cut protection, abrasion resistance and comfort
$112.30 - $195.74
Specifications
Cut Resistance Rating Level 4
For Use With (Application) appliance manufacturing, automotive, general carpentry, general industry, Glass, masonry/tile, Metal fabrication, Metalworking, mining, plumbers, textiles
Glove Cuff Style Knit
Weight Medium weight
View More Specs
Products
Catalog Number Mfr. No. Color Material Size Price Quantity    
19-920-1769
ansell™
707616245732
Dozen of 12 Each for $112.30
Case of 6 Dozen for $673.80
19-920-1770
ansell™
707617245708
Dozen of 12 Each for $112.30
Case of 6 Dozen for $673.80
19-920-1772
ansell™
707619245710
Green Kevlar™ Fiber 9 Dozen of 12 Each for $112.30
Case of 6 Dozen for $673.80
19-920-1773
ansell™
7076110245711
Dozen of 12 Each for $112.30
Case of 6 Dozen for $673.80
19-920-1775
ansell™
707658245714
Mint green Kevlar™ Fiber, Leather 8 Dozen of 12 Each for $195.74
Case of 3 Dozen for $587.22
19-920-1771
ansell™
707618245709
Mint green Kevlar™ Fiber 8 Dozen of 12 Each for $112.30
Case of 6 Dozen for $673.80
19-920-1776
ansell edmont industrial
707659245715
Mint green Kevlar™ Fiber, Leather 9 Dozen of 12 Pair for $195.74
Case of 3 Dozen for $587.22
19-920-1778
ansell™
7076511245717
Mint green Kevlar™ Fiber, Leather 11 Dozen of 12 Each for $195.74
Case of 3 Dozen for $587.22
19-920-1777
ansell edmont industrial
7076510245716
Mint green Kevlar™ Fiber, Leather 10 Dozen of 12 Pair for $195.74
Case of 3 Dozen for $587.22
19-920-1781
ansell™
708609245725
Dozen of 12 Each for $115.78
Case of 6 Dozen for $694.68
Description & Specifications
Specifications
Cut Resistance Rating Level 4
For Use With (Application) appliance manufacturing, automotive, general carpentry, general industry, Glass, masonry/tile, Metal fabrication, Metalworking, mining, plumbers, textiles
Glove Cuff Style Knit
Weight Medium weight
Revolutionary engineered yarns provide increased cut protection without the bulk. Ideal for work environments where sharp parts and materials are handled.
  • Blended fiber, lightweight construction provides all-day comfort
  • Contour design offers flexibility for reduced hand fatigue
  • Medium weight, 10-gauge yarn with 100% Kevlar™ reinforced thumb crotch
ANSI™ Level 4 cut protection