Please login to your online account to display your discounted pricing
Ansell™ Touch N Tuff™ Disposable Nitrile Gloves
Proven splash resistance against hazardous chemicals. Ansell Edmont Touch N Tuff™ Disposable Nitrile Gloves provide a comfortable fit and minimize hand fatigue.
$12.28 - $12.93
Specifications
Color Green
Material Nitrile
Disposable Yes
Hand Ambidextrous
Thickness 5mil
View More Specs
Products
Catalog Number Mfr. No. Certifications Thickness Length (Metric) Size Price Quantity    
18-999-266B
ansell™
925008
FDA, 21 CFR 177-2600 5mil 24cm Medium Pack of 100 Each for $12.92
Case of 10 Pack for $123.60
18-999-266D
ansell™
9250010
FDA, 21 CFR 177-2600 5mil 24cm X-Large Pack of 100 Each for $12.92
Case of 10 Pack for $123.60
18-999-266C
ansell™
925009
FDA, 21 CFR 177-2600 5mil 24cm Large Pack of 100 Each for $12.92
Case of 10 Pack for $123.60
18-999-266A
ansell™
925007
FDA, 21 CFR 177-2600 5mil 24cm Small Pack of 100 Each for $12.92
Case of 10 Pack for $123.60
18-999-2748A
ansell™
926007
FDA, 21 CFR 177-2600 5mil 24cm Small Pack of 100 Each for $12.93
Case of 10 Pack for $123.70
18-999-2748B
ansell™
926008
FDA, 21 CFR 177-2600 5mil 24cm Medium Pack of 100 Each for $12.28
Case of 10 Pack for $117.50
18-999-2748C
ansell™
926009
FDA, 21 CFR 177-2600 5mil 24cm Large Pack of 100 Each for $12.93
Case of 10 Pack for $123.70
18-999-2748D
ansell™
9260010
FDA, 21 CFR 177-2600 5mil 24cm X-Large Pack of 100 Each for $12.93
Case of 10 Pack for $123.70
Description & Specifications
Specifications
Color Green
Material Nitrile
Disposable Yes
Hand Ambidextrous
Thickness 5mil
Offers exceptional resistance to nuisance punctures, nicks, cuts and abrasions. Splash protection against irritating chemicals.
  • Made from 100% nitrile for superior elasticity
  • Comfortable thin mil construction provides good chemical resistance
  • Rolled cuffs for easy donning and doffing
  • Green
Not recommended for Acetone, Benzaldehyde, Chlorobenzene, EDC, DMAC, DMF, Ethyl Acetate, Furfural, Methylene Bromide, MEK, Morpholine, NMP, Iodomethane, Nitric Acid -70%, (Concentrated), itric Acid, Red Fuming, Nitrobenzene, 1-Nitropropane, 2-Nitropropane, Phenol, 90%, THF, Pyridine, Sulfuric Acid, 95-98% (Concentrated) and THF