Please login to your online account to display your discounted pricing
Ansell™ Sol-Vex™ Nitrile Gloves
Resistant to chemicals—will not weaken or swell in many solvents, caustics or animal fats. Ansell™ Sol-Vex™ Nitrile Gloves are ideal for numerous applications involving chemical processing.
Manufacturer: ansell™  371756
 View more versions of this product
Catalog No. 11-391-139A
  • / Pair
  • / Case of 144 Pair
Description & Specifications
Specifications
Certifications 21 CFR 177-2600, FDA
Glove Cuff Style Straight
For Use With (Application) chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
Length (English) 13 in.
Length (Metric) 33cm
Lining Flock
Material Nitrile
Palm Type Contoured
Size 6
Thickness 15mil
Color Green