missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ansell AlphaTec™ 18 in. Solvex™ 37-185 Nitrile Gloves
Heavy-duty, elbow length nitrile chemical resistant gloves suitable for a wide range of applications.
Supplier: Ansell 3718511090
Description
Heavy-duty, elbow length nitrile chemical resistant gloves suitable for a wide range of applications.
- High performance nitrile compound
- Combination of chemical resistance and strength ,for optimal results in wet or dry environments
- Snag, puncture and abrasion protection compared with rubber or neoprene gloves
- High levels of flexibility, comfort and dexterity
- Suitable for heavy-duty cleaning applications
- Complies with FDA food handling requirements (21 CFR 177.2600)
Specifications
ANSI 4 | |
CE 0493, EN 420:2003 + A1:2009, Category III | |
Green | |
Nitrile | |
22 mil | |
EN 388:4102X, EN 388:4102 | |
Type A | |
Bacteria, Fungi only | |
Gauntlet | |
Unlined | |
37-185 9 117300 | |
18 in. |
FDA 21 CFR 177.2600 | |
Yes | |
Yes | |
9 | |
0.55 mm | |
No | |
AGJKLPT | |
Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work | |
Sandpatch | |
ISO 18889 | |
AlphaTec™ Solvex™ 37-185 | |
45.7 cm |