missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ansell AlphaTec™ 15 in. Solvex™ 37-165 Nitrile Gloves
Heavy-duty 22 mil nitrile gloves offer versatile chemical protection that performs across a diverse range of applications.
Supplier: Ansell 3716511070
Description
Heavy-duty 22 mil nitrile gloves offer versatile chemical protection that performs across a diverse range of applications.
- Nitrile construction protects against abrasions, cuts, snags and punctures
- Curved finger design and contoured palm help reduce fatigue
- Full cut for easy donning and doffing
- Does not contain protein allergens
- No inner cotton flocking, reducing risk of lint contamination
- Sandpatch finish gives the glove a smooth surface
- Fully reusable, with an unequaled abrasion resistance
- Complies with FDA food handling requirements (21 CFR 177.2600)
Specifications
ANSI 5 | |
CE 0493, EN 420:2003 + A1:2009, Category III | |
Green | |
Nitrile | |
22 mil | |
EN 388:4102, EN 388:4102X | |
Type A | |
Bacteria, Fungi only | |
Gauntlet | |
Unlined | |
AlphaTec™ Solvex™ 37-165 | |
38 cm |
FDA 21 CFR 177.2600 | |
Yes | |
Yes | |
7 | |
0.55 mm | |
No | |
AGJKLPT | |
Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work | |
Sandpatch | |
37-165 7 117207 | |
15 in. |