Please login to your online account to display your discounted pricing
Ansell™ Sol-Vex™ Nitrile Gloves
Resistant to chemicals—will not weaken or swell in many solvents, caustics or animal fats. Ansell™ Sol-Vex™ Nitrile Gloves are ideal for numerous applications involving chemical processing.
Manufacturer: ansell™  3718510
 View more versions of this product
Catalog No. 11-391-140D
  • / Pair
  • / Dozen of 12 Pair
Description & Specifications
Specifications
Certifications 21 CFR 177-2600, FDA
Glove Cuff Style Straight
For Use With (Application) chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
Length (English) 18 in.
Length (Metric) 45.7cm
Lining None
Material Nitrile
Palm Type Contoured
Size 10
Thickness 22mil
Color Green