Ansell™ Sol-Knit™ Gloves
Offers chemical resistance of nitrile with the strength and comfort of cotton
Manufacturer:  Ansell™ 3912410
 View more versions of this product
Catalog No. 11-394-39D
Specifications Description & Specifications
Specifications
Rough
Chemical plants and transportation, chemical processing, oil refining and petrochemicals, cleaning and de-greasing, exposure to chemicals, solvents, hazardous liquids, PVC cements or standing water, janitorial and maintenance work
Nitrile
21 CFR 177-2600, FDA
14 in.
Gauntlet
Cotton
10
Green
35.5cm