Please login to your online account to display your discounted pricing
Ansell™ Sol-Knit™ Gloves
Offers chemical resistance of nitrile with the strength and comfort of cotton
Manufacturer: ansell™  391248
 View more versions of this product
Catalog No. 11-394-39B
  • / Pair
  • / Case of 72 Pair
Description & Specifications
Specifications
Certifications 21 CFR 177-2600, FDA
For Use With (Application) Chemical plants and transportation, chemical processing, oil refining and petrochemicals, cleaning and de-greasing, exposure to chemicals, solvents, hazardous liquids, PVC cements or standing water, janitorial and maintenance work
Lining Cotton
Glove Cuff Style Gauntlet
Length (English) 14 in.
Length (Metric) 35.5cm
Material Nitrile
Size 8
Coating Material Rough
Color Green