Please login to your online account to display your discounted pricing
Ansell™ Sol-Knit™ Gloves
Reinforced nitrile with comfortable reinforced cotton liner
Manufacturer: ansell™  391229
 View more versions of this product
Catalog No. 11-394-38C
  • / Pair
  • / Case of 72 Pair
Description & Specifications
Specifications
Length (English) 12 in.
Size 9
Length (Metric) 30.5cm
Color Green
Certifications 21 CFR 177-2600, FDA
For Use With (Application) chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
Glove Cuff Style Gauntlet
Material Nitrile
Coating Material Rough
Lining Cotton