Please login to your online account to display your discounted pricing
Ansell™ Polar Bear™ Plus 35 Gloves
Made with high-performance Spectra™ fibers
$20.09 - $232.96
Products
Catalog Number Mfr. No. Price Quantity    
19-058965
ansell™
740357*
Each for $20.09
19-058952
ansell™
740358*
Each for $20.09
Case of 12 Each for $220.08
19-059878
ansell™
740356
Case of 12 Each for $232.96
Description & Specifications
Lightweight, soft, and exceptionally flexible. Ideal for use in meatpacking, food processing, materials handling and numerous other applications requiring an exceptionally high level of protection from cuts.
  • Combines high-performance fiber with other ultratough filaments
  • Provides outstanding protection from cuts and abrasion
  • Can be worn comfortably for extended periods
  • Machine launderable and hand-to-hand reversible, so no pairing is required after laundering
  • Rib-knitted wrist and high-visibility color-coded wrist trim
  • Color: White
Sold by the glove.