Please login to your online account to display your discounted pricing
Ansell™ Petroflex™ PVC-Coated Gauntlets
Rugged, rough finish adds cut and abrasion resistance and excellent wet grip
$3.56 - $3.56
Products
Catalog Number Mfr. No. Price Quantity    
29-635-8652
ansell™
12-212 10 204854
Pair for $3.56
Description & Specifications
For applications where gloves become excessively contaminated or where glove controls are difficult to administer. Ideal for use in the petrochemical industry, machining, construction, forestry, or mining. Also suited for handling equipment and abrasive materials coated with oil, grease, acids, and solvents.
  • Jersey lined for cushioning and moderate insulation; two-piece lining eliminates seams from work areas to prevent irritation and premature wear-through
  • Curved, preflexed fingers and wing thumb give extra comfort and flexibility; two-piece lining eliminates seams from work areas to prevent irritation and premature wear-through
  • Length: 12 in. (30.5cm)
  • Large size only