Please login to your online account to display your discounted pricing
Ansell Perry Original Style 42 Latex Surgical Gloves
$52.88 - $52.88
Specifications
Certifications ASTM D 1076, ISO9001, ISO13485, EN 46001, ASTM D 3577, Type 1
Coating Material Satin smooth
Color White
Material Natural Rubber
Sterile Yes
View More Specs
Products
Catalog Number Mfr. No. Width (English) Length (English) Size Price Quantity    
11-130-3
ansell edmont industrial
5711104
3.7 in. 11.7 in. 7.5 Pack of 50 Each for $52.88
Case of 4 Pack for $211.52
Description & Specifications
Specifications
Certifications ASTM D 1076, ISO9001, ISO13485, EN 46001, ASTM D 3577, Type 1
Coating Material Satin smooth
Color White
Material Natural Rubber
Sterile Yes
Disposable Yes
Glove Cuff Style Beaded
Hand Hand-specific
Powdered Yes
Thickness 8.3mil