Please login to your online account to display your discounted pricing
Ansell™ Omni™ Gloves
Cotton flock lined for maximum user comfort
Manufacturer: ansell™  27610
 View more versions of this product
 Alert
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 11-394-90D
  • / Dozen of 12 Pair
  • / Case of 12 Dozen
Description & Specifications
Specifications
Certifications 21 CFR 177-2600, FDA
For Use With (Application) blend of Neoprene and natural Latex resists tearing, puncturing, abrasions and various chemicals, including Caustics, animal fats, Alcohols, Acids, Salts, Detergents and Petroleum based Solvents
Glove Cuff Style Embossed Pebble
Color Light green
Thickness 20mil
Material Neoprene, Natural Rubber Latex
Size 10
Length (Metric) 30.5cm
Lining Cotton Flock
Length (English) 12 in.