Ansell AlphaTec™ 87-276 Natural Rubber/Neoprene Omni Gloves
Medium weight, cotton flock lined gloves provide chemical, snag, puncture and abrasion resistance.
$5.41 - $30.02
Specifications
Certifications/Compliance 21 CFR 177.2600
Certifications/Compliance (Europe) CE 0493
Chlorinated Yes
Color Green
Latex Free No
View More Specs
Alert:
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Products 4
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
11-394-90A
View Documents Promotion Details
Ansell
0027611070
7
Dozen of 12 Pair for $6.97
Case of 12 Dozen for $83.64
11-394-90B
View Documents Promotion Details
Ansell
0027611080
8
Dozen of 12 Pair for $5.41
Case of 12 Dozen for $64.92
11-394-90C
View Documents Promotion Details
Ansell
0027611090
9
Dozen of 12 Pair for $27.71
Case of 12 Dozen for $317.88
11-394-90D
View Documents Promotion Details
Ansell
0027611100
10
Dozen of 12 Pair for $30.02
Case of 12 Dozen for $344.40
Description
Description
Blend of neoprene and natural latex resists tearing, puncturing, abrasions and various chemicals, including caustics, animal fats, alcohols, acids, salts, detergents and petroleum-based solvents
  • Ideal for food processing, automotive manufacturing, aircraft assembly, battery manufacturing, chemical plants, pesticide manufacturing and plant maintenance
  • Embossed “pebble” pattern enables a sure grip on wet or dry surfaces
  • Length: 12 in. (30.5 cm)
  • Thickness: 20 mil
Specifications
Specifications
21 CFR 177.2600
Yes
No
20 mil
EN 388:1010A, EN 388:2110
KLP
Pinked
Cotton Flock
AlphaTec™ 87-276
30.5 cm
CE 0493
Green
Latex, Neoprene
0.50 mm
Type B
Bacteria, Fungi only
Pebble Embossed
Lined
12 in.
SDS
missing translation for 'documents'
missing translation for 'documents'