Please login to your online account to display your discounted pricing

Ansell™ Micro-Touch™ Plus Latex Exam Gloves

$207.30 - $223.25

Specifications

Color Cream
Length (English) 9.5 in.
Length (Metric) 241mm
Material Latex
Sterility Non-sterile
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

17-200-825

 
ansell edmont industrial
6015301
Small Case of 1000 Each for $223.25

17-200-826

 
ansell edmont industrial
6015302
Medium Case of 1000 Each for $207.30
Description & Specifications

Specifications

Color Cream
Length (English) 9.5 in.
Length (Metric) 241mm
Material Latex
Sterility Non-sterile
Glove Cuff Style Beaded/Rolled

510K approved for medical use