Please login to your online account to display your discounted pricing

Ansell™ Latex Canners and Handlers Gloves


Unlined industrial latex gloves

Manufacturer: ansell™  883928193923

 View more versions of this product

 Alert

These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Catalog No. 11-391-135B

  • / Dozen of 12 Pair
  • / Case of 12 Dozen
 

Description & Specifications

Specifications

Certifications 21 CFR 177-2600, FDA
For Use With (Application) canning and food processing, small parts assembly, pharmaceutical manufacturing and plant maintenance, resist Acids, Detergents, vegetable Oil, Salts, Caustics and Alcohols
Glove Cuff Style Pinked
Exterior Finish Pebble embossed
Size 8
Material Natural Rubber
Color Natural
Thickness 20mil
Powdered No
Length (Metric) 30.5cm
Sterile No
Length (English) 12 in.