Please login to your online account to display your discounted pricing
Ansell™ Latex Canners and Handlers Gloves
Unlined industrial latex gloves
Manufacturer: ansell™  883927193921
 View more versions of this product
 Alert
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 11-391-135A
  • / Dozen of 12 Pair
  • / Case of 12 Dozen
Description & Specifications
Specifications
Size 7
Powdered Powder-free
Color Natural
Material Rubber, natural
Thickness (English) 20 mil
Length (English) 12 in.
Length (Metric) 30.5cm
Sterility Non-sterile
Certifications 21 CFR 177-2600, FDA
For Use With (Application) canning and food processing, small parts assembly, pharmaceutical manufacturing and plant maintenance, resist Acids, Detergents, vegetable Oil, Salts, Caustics and Alcohols
Glove Cuff Style Pinked
Exterior Finish Pebble embossed