Please login to your online account to display your discounted pricing

Ansell™ KSR™ Vinyl-Coated Gloves


Light, flexible, and comfortable

Manufacturer: ansell™  225159**

 View more versions of this product

Catalog No. 11-391-121B

  • / Pair
  • / Case of 144 Pair
 

Description & Specifications

Specifications

Size 9
Length (English) 9.64 in.
Length (Metric) 24.5cm
Color Tan
For Use With (Application) assembling, inspecting and packing small or delicate parts
Glove Cuff Style Slip-on
Cut Resistance Rating Level 1
Exterior Finish Textured
Material Vinyl
Cleaning Procedure Launderable