Please login to your online account to display your discounted pricing

Ansell™ Hynit™ Gloves


Gloves outwear 8 oz. cotton as much as ten to one; goatskin, three to one; and light leather, four to one

Manufacturer: ansell™  321059

 View more versions of this product

Catalog No. 11-391-131B

  • / Pair
  • / Case of 144 Pair
 

Description & Specifications

Specifications

Glove Cuff Style Slip-on
For Use With (Application) general plant use, assembly inspection, receiving and light to medium fabrication and materials handling
Exterior Finish Smooth
Size 9
Material Nitrile, Impregnated Fabric
Color Blue
Glove Cuff Color Brown
Cleaning Procedure Machine washable
Coated Area Entire hand