Ansell™ HyFlex™ Ultralight Weight Multi-purpose Gloves
Gloves feature ZONZ™ Comfort Fit technology
Manufacturer:  Ansell™ 118188111715
 View more versions of this product
Catalog No. 19-148-412
Description
Description
  • Under abrasive conditions, ultra-thin FORTIX™ nitrile foam coating (patent pending) lasts up to three times longer than comparable coatings, extending glove life and reducing total cost of ownership
  • FORTIX coating provides enhanced dry grip and is 20% more breathable than earlier formulations, for cooler, drier hands and less sweat
  • Seamless 18-gauge nylon-spandex liner has a barehand feel, offering breathability, comfort and enhanced fingertip sensitivity for precision tasks
  • Ansell ZONZ Comfort Fit technology matches natural hand contours for improved support, breathability and range of movement
  • With EN Abrasion Level 4 , the ultra-lightweight HyFlex11-818 offers ideal protection for a range of general handling and assembly tasks
  • Silicone-free material means no transfer of silicone contaminants to metal parts prior to painting
Specifications
Specifications
Assembly of dry parts, feeding lines, fine mounting of small nuts and bolts, Handling of small/fine parts, Mounting, dismounting parts, Screwing & unscrewing, ultra-fine mechanical applications
8
Dark Blue
200-248mm
Documents