Ansell™ HyFlex™ Oil-Resistant Gloves
Provides optimal control in slippery situations
Manufacturer:  Ansell™ 119207255002
 View more versions of this product
Catalog No. 19-148-501
Description
Description
  • Ansell patent pending Grip Technology coating
  • Nitrile layer with microscopic channels actively transfers oils and other lubricants away from the gripping surface
  • Increases surface contact for enhanced grip and control
  • Prevents oil penetration and reduces the risk of dermatitis and liquid exposure
  • Multi-layer dipping process provides high abrasion resistance and increases wear life
  • Special nylon fine-gauge liner provides softness, comfort and breathability
Specifications
Specifications
Palm, finger
precision assembly and handling of small parts in applications such as electronics, packaging, inspection and use of sensitive instruments
Heatherr Nylon
Blue
Knit
Nitrile
Small
Documents