Please login to your online account to display your discounted pricing
Ansell™ HyFlex™ CR2 Gloves with Dyneema™
Offer excellent abrasion resistance, cut protection, dexterity and comfort
$104.75 - $148.12
Specifications
Coating Material Polyurethane
Color Gray
Cut Resistance Rating Level 2
Material Dyneema™, Lycra™
For Use With (Application) Plastics injection and molding, Glass handling, sheet Metal assembly (even when slightly Oily), handling sharp, small parts
View More Specs
Products
Catalog Number Mfr. No. Hem Color Size Price Quantity    
19-168-55
ansell™
1162711205709
Blue 11 Dozen of 12 Pair for $104.75
Case of 12 Dozen for $1,202.40
19-168-52
ansell™
116278205688
Yellow 8 Dozen of 12 Pair for $148.12
Case of 12 Dozen for $1,700.16
19-168-53
ansell™
116279205689
Brown 9 Dozen of 12 Pair for $148.12
Case of 12 Dozen for $1,700.16
19-168-50
ansell™
116276205686
Purple 6 Dozen of 12 Pair for $148.12
Case of 12 Dozen for $1,700.16
19-168-51
ansell™
116277205687
Red 7 Dozen of 12 Pair for $148.12
Case of 12 Dozen for $1,700.16
19-168-54
ansell™
1162710205690
Black 10 Dozen of 12 Pair for $148.12
Case of 12 Dozen for $1,700.16
Description & Specifications
Specifications
Coating Material Polyurethane
Color Gray
Cut Resistance Rating Level 2
Material Dyneema™, Lycra™
For Use With (Application) Plastics injection and molding, Glass handling, sheet Metal assembly (even when slightly Oily), handling sharp, small parts
Glove Cuff Style Knit
Weight Medium weight
Polyurethane dipped; Dyneema and Lycra™ liner.
  • Palm coating provides dry grip and longer wear
  • Natural fit liner reduces hand fatigue
  • Excellent grip in dry environments
  • Effective control in minimally oily environments
  • Breathable liner increases worker acceptance and safety compliance
  • Machine launderable
ANSI™ Level 2 cut protection.