Ansell™ HyFlex™ CR Plus Foam-Nitrile-Dipped Work Gloves
Kevlar™ Stretch Armor construction combines superior cut resistance and outstanding dexterity
Manufacturer:  Ansell™ 1150111070
 View more versions of this product
Catalog No. 19-130-11898B
Description
Description
Applications include sheet metal and automotive assembly as well as glass and small parts handling.
  • Intercept Technology™ yarn — developed jointly by Ansell and DuPont™ — incorporates new Kevlar Stretch Armor Technology to produce a lightweight, flexible knit with great cut resistance
  • Gloves feature impressive tactile sensitivity and comfort
  • Nitrile coating on palm, thumb, and fingertips offers excellent gripping in oily conditions
Compliance
ANSI™ Level 4 cut protection.
Specifications
Specifications
Palm
Metal fabrication, body assembly, press and stamping, welding Glass manufacturing, Plastics industry
Dupont™ Kevlar™ aramid fibers
Blue,Gray
20.3 to 25.5 cm
Elastic knit
Foam Nitrile
7
7.99 to 10 in.