Ansell HyFlex™ 11-501 Aramid Fiber Stretch Armor Gloves, Nitrile Coated
Ergonomic design reduces hand fatigue and provides a fit that mimics bare hands. Nitrile foam coating enhances tactile sensitivity and grip capabilities in dry or light oil conditions.
Supplier:  Ansell 1150111060
 View more versions of this product
Catalog No. 19-130-11898A
Description
Description
  • Intercept Technology™ raises the protection & dexterity standards for cut-resistant gloves
  • Aramid fiber provides an additional level of protection
  • Ergonomic design provides bare-hand closeness and reduces hand fatigue
  • Nitrile foam coating enhances grip capabilities and tactile sensitivity
Specifications
Specifications
ANSI 3
Blue
Specific
Cut-resistant Gloves
Automotive Repairs, Machinery/Equipment Operation, Metal Fabrication, and Chemical Handling
11-501
Nitrile
ANSI A5
Aramid Fiber
6
Knit