Ansell™ HyFlex™ CR Gloves
For a sure grip in light oil applications; hides soil
Manufacturer: Ansell™ 1150011060
Description
Provide excellent hand protection in assembly and maintenance operations involving handling of sharp-edged and pointed tools and material.
- Cut-resistant stretch Kevlar™ liner provides comfortable fit and protection from nuisance nicks and abrasion
- Black foam nitrile coating on palm and fingertips channels oil away from glove surface Extended cuff for extra cut protection around wrist
- Yellow
Compliance
ANSI™ Level 2 cut protection.
Specifications
Knit | |
handling and assembly of small, sharp, and/or pointed parts, handling wire, sheet Metal assembly | |
6 | |
Black,Yellow |
Purple | |
Dupont™ Kevlar™ Aramid Fiber | |
Medium weight |