missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ansell™ HyFlex™ CR Gloves

For a sure grip in light oil applications; hides soil

$91.94 - $99.39

Specifications

Color Black,Yellow
Material Dupont™ Kevlar™ Aramid Fiber
For Use With (Application) handling and assembly of small, sharp, and/or pointed parts, handling wire, sheet Metal assembly
Glove Cuff Style Knit
Weight Medium weight
View More Specs

Products 4
Catalog Number Mfr. No. Size Hem Color Price Quantity  
Catalog Number Mfr. No. Size Hem Color Price Quantity  
29-635-7848
View Documents
Ansell™
1150011060
6 Purple
Dozen of 12 Pair for $99.39
Case of 12 Dozen for $1,157.64
 
19-152-154
View Documents
Ansell™
1150011100
10 Black
Dozen of 12 Pair for $97.43
Case of 12 Dozen for $1,134.84
 
19152148
View Documents
Ansell™
1150011110
11 Blue
Dozen of 12 Pair for $91.94
Case of 12 Dozen for $1,055.28
 
19-152-151
View Documents
Ansell™
1150011070
7 Red
Dozen of 12 Each for $97.43
Case of 12 Dozen for $1,134.84
 
Description

Description

Provide excellent hand protection in assembly and maintenance operations involving handling of sharp-edged and pointed tools and material.

  • Cut-resistant stretch Kevlar™ liner provides comfortable fit and protection from nuisance nicks and abrasion
  • Black foam nitrile coating on palm and fingertips channels oil away from glove surface Extended cuff for extra cut protection around wrist
  • Yellow

Compliance

ANSI™ Level 2 cut protection.

Specifications

Specifications

Black,Yellow
handling and assembly of small, sharp, and/or pointed parts, handling wire, sheet Metal assembly
Medium weight
Dupont™ Kevlar™ Aramid Fiber
Knit
SDS