Please login to your online account to display your discounted pricing

Ansell™ Hycron™ Nitrile-Coated Gloves


Soft and comfortable

Manufacturer: ansell™  276008

 View more versions of this product

Catalog No. 11-391-126A

  • / Pair
  • / Case of 144 Pair
 

Description & Specifications

Specifications

Glove Cuff Style Knit
For Use With (Application) casting and core manufacturing, handling Metal cable, bricks and blocks
Size 9
Coating Material Nitrile
Color Blue
Cut Resistance Rating Level 1