Please login to your online account to display your discounted pricing

Ansell™ Hycron™ Nitrile-Coated Gloves

Soft and comfortable

$4.21 - $4.37

Specifications

Coating Material Nitrile
Color Blue
Size 10
For Use With (Application) casting and core manufacturing, handling Metal cable, bricks and blocks
Glove Cuff Style Knit
View More Specs
Products
Catalog Number Mfr. No. Cut Resistance Rating Price Quantity    

11-391-126A

 
ansell™
276008
Level 1 Pair for $4.21
Case of 144 Pair for $540.00

11-391-127B

 
ansell™
2760210
Level 2 Pair for $4.31
Case of 144 Pair for $554.40

11-391-128B

 
ansell™
2760710
Level 2 Pair for $4.37
Case of 144 Pair for $561.60

11-391-126B

 
ansell™
2760010
Level 1 Pair for $4.21
Case of 144 Pair for $540.00
Description & Specifications

Specifications

Coating Material Nitrile
Color Blue
Size 10
For Use With (Application) casting and core manufacturing, handling Metal cable, bricks and blocks
Glove Cuff Style Knit

Available in three styles: nitrile-coated palm with knitwrist, full nitrile coating with knitwrist, and nitrile-coated palm with safety cuff. Nitrile coating ensures excellent handling of rough, abrasive materials while providing superior resistance to cuts, snags, punctures, abrasion, grease, and oil.

  • Two-piece jersey lining eliminates seams from working areas to prevent irritation and premature wear-through
  • Curved, preflexed fingers and wing thumb offer maximum worker comfort
  • Ideal for handling heavy-duty castings, cores, fabricated metal, concrete block, brick, tile, lumber, drywall, and plywood