Please login to your online account to display your discounted pricing
Ansell™ FL200 Flock-Lined Latex Gloves
Superior tensile strength and sensitivity
$1.30 - $15.64
Specifications
Certifications 21 CFR 177-2600, FDA
Length (English) 12 in.
Length (Metric) 30.5cm
Material Natural Rubber
Sterile No
View More Specs
 Alert
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Products
Catalog Number Mfr. No. Color Exterior Finish Size Price Quantity    
11-394-7C
ansell™
2989
Lemon yellow Fishscale 9 Dozen of 12 Pair for $15.24
Case of 12 Dozen for $174.84
11-394-7A
ansell™
2987
Lemon yellow Fishscale 7 Dozen of 12 Pair for $15.24
Case of 12 Dozen for $174.84
11-394-7B
ansell™
2988
Lemon yellow Fishscale 8 Dozen of 12 Pair for $15.24
Case of 12 Dozen for $174.84
11-394-7D
ansell™
29810
Lemon yellow Fishscale 10 Dozen of 12 Pair for $15.64
Case of 12 Dozen for $179.52
19-014504
ansell™
2978
Pair for $1.30
Case of 144 Pair for $155.52
19-014506
ansell™
2979
Pair for $1.30
Case of 144 Pair for $155.52
19-014507
ansell edmont industrial
29710
Pair for $1.33
Case of 144 Pair for $159.84
Description & Specifications
Specifications
Certifications 21 CFR 177-2600, FDA
Length (English) 12 in.
Length (Metric) 30.5cm
Material Natural Rubber
Sterile No
For Use With (Application) chemical plants, janitorial services, automotive assembly, plant maintenance, agriculture, Petroleum refineries, canning and pharmaceutical manufacturing, resist Acids, Detergents, Salts, Caustics, animal fats and Alcohols
Glove Cuff Style Pinked
Lining Flock
Powdered No
Thickness 20mil
All styles are ideal for chemical plants, janitorial services, automotive assembly, plant maintenance, agriculture, petroleum refineries, canning and pharmaceutical manufacturing. Resist acids, detergents, salts, caustics, animal fats and alcohols.
  • Made from natural latex for excellent abrasion, puncture and tear resistance
  • Two styles available with pattern embossing for secure grip of wet or dry objects
  • Length: 12 in.(30.5cm)
  • Thickness: 20mil
  • Available in yellow or blue
Component materials of Styles 254 and 255 comply with 21CFR177.2600 for food contact.