Please login to your online account to display your discounted pricing

Ansell™ Crusader™ Flex Gloves

Non-woven felt insulation and a full-length heat barrier designed for intermittent handling of objects up to 400°F. Ansell™ Crusader™ Flex Gloves can be used in industries such as food service, and glass and metal fabrication.

$16.96 - $17.10

Specifications

Cleaning Procedure Launderable
Coated Area Fully coated
Color Light gray
Cut Resistance Rating ANSI 2
Exterior Finish Nitrile
View More Specs
Products
Catalog Number Mfr. No. Length (English) Material Size Price Quantity    

19-019857

 
ansell™
4247410
14 in. Back of hand heat insulation 10 (Large) Pair for $16.96
Case of 72 Pair for $1,135.44

19-019856

 
ansell™
424749
14 in. Back of hand heat insulation 9 (Medium) Pair for $17.10
Case of 72 Pair for $1,144.08
Description & Specifications

Specifications

Cleaning Procedure Launderable
Coated Area Fully coated
Color Light gray
Cut Resistance Rating ANSI 2
Exterior Finish Nitrile
For Use With (Application) Food service, glass and metal fabrication
Liner Material Non-woven felt

  • Front-and-back heat-insulated styles
  • Provides protection from radiant heat and intermittent back-of-hand contact with hot objects
  • Provides excellent grip when handling dry or oily surfaces
  • Gauntlet slip-on cuff
  • Contains no asbestos
  • Length: 10 in. (25.4cm)

 

Not a replacement for silicate gloves. Designed for intermittent handling of objects up to 400°F (204°C).