Please login to your online account to display your discounted pricing
Ansell™ ChemTek™ Viton/Butyl Chemical Handling Gloves
For aggressive chemical handling
$1,167.24 - $1,167.24
Products
Catalog Number Mfr. No. Price Quantity    
19-162-033
ansell edmont industrial
3861210103212
Dozen of 12 Each for $1,167.24
Case of 3 Dozen for $3,501.72
Description & Specifications
  • Offers an extra level of protection in applications involving aromatic chemicals and chlorinated solvents
  • Viton layer over butyl means lower-cost glove for aggressive chemical exposure
  • Delivers unique balance of ergonomic design and tactile sensitivity with optimum chemical splash protection
  • Smooth grip
  • Length: 21 in. (30.4cm)
Each pair is packed with an individual informational insert so critical information is always at your fingertips.