Please login to your online account to display your discounted pricing
Ansell™ ChemTek™ Butyl Chemical Handling Gloves
Provides the best resistance to the most aggressive chemicals. Ansell Edmont™ ChemTek™ Butyl Chemical Handling Gloves protect hands without compromising dexterity or comfort.
$464.99 - $464.99
Specifications
Color Black
Material Butyl
Exterior Finish Rough
For Use With (Application) chemical plants and transportation, chemical processing, handling Acids, Alkalies, Salts and Ketones, handling specialty chemicals, Hazmat work
Glove Cuff Style Beaded
View More Specs
Products
Catalog Number Mfr. No. Length (English) Length (Metric) Size Price Quantity    
19-161-432C
ansell™
385149
12 in. 30.4cm 9 Dozen of 12 Pair for $464.99
Case of 3 Dozen for $1,394.97
Description & Specifications
Specifications
Color Black
Material Butyl
Exterior Finish Rough
For Use With (Application) chemical plants and transportation, chemical processing, handling Acids, Alkalies, Salts and Ketones, handling specialty chemicals, Hazmat work
Glove Cuff Style Beaded
Thickness 14mil
  • The ChemTek™ butyl 38-514 offers superior protection against aldehydes, ketones and esters as well as concentrated mineral acids
  • ChemTek™ gloves are surprisingly flexible and provide excellent dexterity. Designed for comfort – with a natural, curved ergonomic shape and soft feel – they offer easy donning and good grip
  • The absence of latex proteins means there’s no risk of latex allergies
Each pair is packed with an individual informational insert so critical information is always at your fingertips.