Please login to your online account to display your discounted pricing
Ansell™ Black Bear™ Gloves
$17.83 - $17.83
Products
Catalog Number Mfr. No. Price Quantity    
19-056257
ansell edmont industrial
744019
Each for $17.83
Case of 12 Each for $195.36
Description & Specifications
Similar to Ansell™Polar Bear™ Supreme Gloves (Series 19-053), but black. Ideal for handling nonfood items, such as in blade changing; machine maintenance; and glass, ceramics, and sheet metal handling. Full fingered. Style: PBS401. Sold by the glove.