Please login to your online account to display your discounted pricing
Ansell™ AlphaTEC™ Gloves
Provides dexterity with resistance to cuts, punctures, and abrasions as well as petrochemicals, oils, greases, pesticides, and other agricultural chemicals
$91.78 - $111.58
Specifications
Color Red with black coating
For Use With (Application) Chemical plants and transportation, chemical processing; oil refining and petrochemicals; cleaning and de-greasing; exposure to chemicals, solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
View More Specs
Products
Catalog Number Mfr. No. Length (English) Length (Metric) Size Price Quantity    
19-161-447
ansell™
585309
12 in. 30.5cm 9 Dozen of 12 Pair for $91.78
Case of 6 Dozen for $550.68
19-161-446
ansell™
585308
12 in. 30.5cm 8 Dozen of 12 Pair for $91.78
Case of 6 Dozen for $550.68
19-161-451
ansell™
58- 535 10 219009
14 in. 35.5cm 10 Dozen of 12 Pair for $105.05
Case of 6 Dozen for $630.30
19-161-448
ansell™
58-530 10 219004
12 in. 30.5cm 10 Dozen of 12 Pair for $91.78
Case of 6 Dozen for $550.68
19-161-450
ansell™
585359
14 in. 35.5cm 9 Dozen of 12 Pair for $91.78
Case of 6 Dozen for $550.68
19-161-449
ansell™
585358
14 in. 35.5cm 8 Dozen of 12 Pair for $109.87
Case of 6 Dozen for $659.22
29-636-1634
ansell™
5853511
14 in. 35.5cm 11 Dozen of 12 Each for $111.58
Description & Specifications
Specifications
Color Red with black coating
For Use With (Application) Chemical plants and transportation, chemical processing; oil refining and petrochemicals; cleaning and de-greasing; exposure to chemicals, solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
  • Chemical resistant
  • Material fully coated nitrile gloves
  • Thickness: 11 mil
  • Embossed grip surface for superior wet and oil grip
  • Red with black coating
  • Comfortable design reduces fatigue
Chemical handling, agriculture, automotive/OEM, mining, aerospace, printing