Please login to your online account to display your discounted pricing
Ansell™ AccuTech™ Ambi UltraGrip Latex Gloves
An outstanding combination of cleanliness and safe handling.
$160.54 - $160.54
Products
Catalog Number Mfr. No. Price Quantity    
19-130-5442A
ansell edmont industrial
91104XS574086
Case of 1000 Pair for $160.54
Description & Specifications
Provide excellent grip in dry or wet applications, due to specific surface treatment as well as textured finish at fingertip level. This enhances dexterity and tactility for intricate applications.
  • Ambidextrous, powder free
  • Designed for critical environments; very low particle and extractable levels
  • Provide exceptional wet or dry grip for safe product handling
  • Easy to don and comfortable for the wearer
  • Ideal for flat panel, electronics, pharmaceutical, and medical device manufacturing applications
  • Also for biotechnologies applications within a clean, but nonsterile environment
  • Can be used as under-gloving in isolators
  • Rolled cuff prevents glove from rolling down
  • Length: 11.5 in. (29.2cm)
  • Thickness: 6mil
Conformance certified. Lot numbers and expiry dates are displayed on all packaging, from master bag to shipping carton. Each lot is supported with a Certificate of Conformance (COC).