Please login to your online account to display your discounted pricing

ANR 94, Tocris™

Adenosine A2A antagonist

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material ANR 94
Assay Percent Range >99%
Molecular Formula C9H13N5O
Formula Weight 207.23
Synonym 8-Ethoxy-9-ethyl-9H-purin-6-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

393710

 
r & d systems
393710
10mg Each for $244.81

393750

 
r & d systems
393750
50mg Each for $997.23
Description & Specifications

Specifications

Formula Weight 207.23
Quantity 10mg
CAS 634924-89-3
Recommended Storage Store at Room Temperature
Chemical Name or Material ANR 94
Assay Percent Range >99%
Molecular Formula C9H13N5O
Synonym 8-Ethoxy-9-ethyl-9H-purin-6-amine
Safety and Handling

Store at Room Temperature