Please login to your online account to display your discounted pricing

Anpirtoline hydrochloride, Tocris


Highly potent 5-HT1B agonist; Also 5-HT3 antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS070310

 View more versions of this product

Catalog No. 070310

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 265.2
Molecular Formula C10H13ClN2S·HCl
Chemical Name or Material Anpirtoline hydrochloride
Synonym 6-Chloro-2-[piperidinyl-4-thio]pyridine hydrochloride
Assay Percent Range >99%
CAS 99201-87-3
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material Anpirtoline hydrochloride
Assay Percent Range >99%
Molecular Formula C10H13ClN2S·HCl
Formula Weight 265.2
Synonym 6-Chloro-2-[piperidinyl-4-thio]pyridine hydrochloride
Accessories

Desiccate at +4°C