Please login to your online account to display your discounted pricing

eBioscience™ Annexin V, Recombinant Protein

Recombinant Protein

$187.05 - $1,825.55

Specifications

Name Annexin V
Recombinant Yes
Conjugate Unlabeled
Purity >89%
Purification Method SDS-gel electrophoresis and reverse phase HPLC
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

509282

 
life technologies
LSBMS3061MG
1mg Each for $1,825.55

509283

 
life technologies
LSBMS306300UG
300μg Each for $912.05

509284

 
life technologies
LSBMS30630UG
30μg Each for $187.05
Description & Specifications

Specifications

Name Annexin V
Recombinant Yes
Conjugate Unlabeled
Purity >89%
Purification Method SDS-gel electrophoresis and reverse phase HPLC
Source E. coli
For Use With (Application) Flow Cytometric Analysis
Molecular Weight 35.8kDa
Storage Requirements The lyophilized protein is stable at 2-8deg;C as indicated on the label. Reconstituted Annexin V should be stored in working aliquots at -20°C.

Annexins are a family of calcium-dependent phospholipid-binding proteins, which bindto phosphatidylserine (PS) to identify apoptotic cells. In healthy cells, PS is predominantlylocated along the cytosolic side of the plasma membrane. Upon initiation of apoptosis,PS loses its asymmetric distribution in the phospholipid bilayer and translocates tothe extracellular membrane, which is detectable with fluorescently labeled AnnexinV. In early stages of apoptosis, the plasma membrane excludes viability dyes such aspropidium iodide (PI) and 7-AAD, therefore cells which display only Annexin V staining(PI/7-AAD negative) are in early stages of apoptosis. During late-stage apoptosis, loss ofcell membrane integrity allows Annexin V binding to cytosolic PS, as well as cell uptakeof PI and 7-AAD. Annexin V staining, paired with 7-AAD or PI is widely used to identifyapoptotic stages by flow cytometry.