missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

eBioscience™ Annexin V-FITC Apoptosis Detection Kit, for in vitro diagnostic use

Annexin V staining, paired with 7-AAD or PI is widely used to identify apoptotic stages by flow cytometry.

$159.50 - $346.55

Specifications

For Use With (Application) Flow Cytometric Analysis
Synonym Lipocortin V, Endonexin II, Calphobindin I
Assay >98%
Conjugate FITC
Formula Weight 35.8 kd
View More Specs

Disclaimers:

For Research Use Only


Includes: Annexin V-FITC protein
Binding Buffer (4x)
Propidium Iodide (20 ug/mL)


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501125437 Life Technologies
LSBMS500FI100CE
100 tests Each for $346.55
501125438 Life Technologies
LSBMS500FI20CE
20 tests Each for $159.50
Specifications

Description

  • Blue Laser
  • Excitation at 488nm
  • Emission at 520nm
Description & Specifications

Specifications

Flow Cytometric Analysis
>98%
35.8 kd
Lipocortin V, Endonexin II, Calphobindin I
FITC